Learn More
Medchemexpress LLC Tetrapentylammonium bromide | 866-97-7 | 98.0% | 378.48 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01105210G
Encompass_Preferred
Tetrapentylammonium bromide is a quaternary ammonium salt consisting of a positively charged tetrapentylammonium cation and a negatively charged bromide anion. It is commonly used as a phase transfer catalyst in organic synthesis reactions to facilitate the transfer of reactants between immiscible phases. It is also used in the production of surfactants, detergents, and in the synthesis of pharmaceuticals. This biochemical reagent can be used as a biological material or organic compound for life science related research.
  • Biochemical assay reagent
  • Quaternary ammonium salt
  • Used as a phase transfer catalyst
  • Used in the production of surfactants and detergents
  • Used in the synthesis of pharmaceuticals
  • Facilitates transfer of reactants between immiscible phases in organic synthesis
Catalog No. 50-004-52470