Learn More
Medchemexpress LLC Tetraphenylporphyrin | 917-23-7 | MFCD00011680 | 99.4% | 614.74 | C44H30N4 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY103656250MG
Encompass_Preferred
Tetraphenylporphyrin is a symmetrically substituted porphyrin used as a structural building block in supramolecular chemistry, photophysical studies, and materials research. Supplied as a solid research reagent, the material is characterized by high purity and accompanied by analytical documentation for laboratory use.
  • High purity suitable for research and synthesis
  • Well-characterized molecular formula and molecular weight
  • Solid form for convenient handling and storage
  • Useful scaffold for supramolecular and photophysical applications
  • Analytical support available (datasheet, COA, SDS)
Catalog No. 50-002-44584