Learn More
Medchemexpress LLC Tetrapropylammonium iodide | 631-40-3 | 313.26 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW009202100G
Encompass_Preferred
Tetrapropylammonium iodide is an organic compound that is commonly used as an ionic liquid and a catalyst. It can function as a solvent in certain chemical reactions and also promotes organic synthesis reactions. Additionally, this compound finds widespread application in batteries, solar cells, and pigments. For research use only, this product is not sold to patients.
  • Used as ionic liquids and catalysts.
  • Can be used as a solvent in chemical reactions.
  • Promotes organic synthesis reactions.
  • Widely used in batteries, solar cells, and pigments.
  • Biochemical reagent for life science-related research.
Catalog No. 50-004-53378