Learn More
Medchemexpress LLC Tetrapropylammonium perruthenate | 114615-82-6 | 351.43 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00780110G
Encompass_Preferred
Tetrapropylammonium perruthenate (TPAP) is an organic compound utilized as a catalyst and oxidizing agent. It is capable of catalyzing oxidation reactions of olefins and aromatic compounds in organic synthesis. Additionally, it finds application in industrial production processes such as plastics, rubber, and textile manufacturing. It is a biochemical reagent for life science research.
  • Utilized as a catalyst and oxidizing agent
  • Capable of catalyzing oxidation reactions of olefins and aromatic compounds in organic synthesis
  • Finds application in industrial production processes such as plastics, rubber, and textile manufacturing
  • Biochemical reagent for life science research
Catalog No. 50-004-46695