Learn More
Strem, An Ascensus Company CAS# 39050-26-5. 1g. Tetrasodium-meso-tetra(4-sulfonatophenyl)porphine dodecahydrate, min. 95%. MFCD00151011
SDP
Supplier:  Strem, An Ascensus Company 0723401G
Encompass
CAS# 39050-26-5. 1g. Tetrasodium-meso-tetra(4-sulfonatophenyl)porphine dodecahydrate, min. 95%. MFCD00151011. Molecular Weight: 1022.87 (1239.11). Molecular Formula: C44H26N4Na4O12S4∙12H2O. Color/form: purple pwdr. Strem# 07-2340. http://www.strem.com/catalog/v/07-2340/
Catalog No. 50-901-09137