Learn More
Medchemexpress LLC Tetrazine-PEG4-biotin | 99.3% | 660.78 | C30H44N8O7S | 5 MG

Supplier: Medchemexpress LLC HY1360535MG
Tetrazine-PEG4-biotin is a cleavable 4-unit polyethylene glycol (PEG) linker that contains a tetrazine moiety and a biotin tag for bioorthogonal conjugation and affinity labeling in research applications. It undergoes rapid inverse electron demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) partners and is supplied as a purple to purplish red solid intended for research use only.
- Cleavable 4-unit PEG linker suitable for antibody-drug conjugate synthesis.
- Facilitates rapid iEDDA click chemistry with TCO-containing molecules.
- Contains a biotin tag for affinity capture or detection.
- High purity reagent (99.3%) supplied as a purple to purplish red solid.
- Recommended storage at -20°C, protect from light; in solvent: -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.