Learn More
Sigma Aldrich Fine Chemicals Biosciences Tetrazole solutin suitabl500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 88185
Encompass
The product is ~0.45M SOLUTIONon of tetrazole in acetonitrile. Tetrazole ring plays significant role in various synthetic and industrial processes.
Catalog No. 50-290-6709