Learn More
Cayman Chemical tetrnor-PGAM 100ug
SDP
Supplier:  Cayman Chemical 13369100 UG
Encompass_Preferred
A dehydration product of tetranor-PGEM that can be measured as a surrogate for tetranor-PGEM levels in urine
Catalog No. 50-295-5701