Learn More
Cayman Chemical tetrnor-PGDM 100ug
SDP
Supplier:  Cayman Chemical 12850100 UG
Encompass_Preferred
A major metabolite of PGD2 found in human and mouse urine at levels of approximately 1.5 and 8.1 ng/mg creatinine, respectively
Catalog No. 50-295-3807