Learn More
Cayman Chemical tetrnor-PGEM 25ug
SDP
Supplier:  Cayman Chemical 1484025 UG
Encompass_Preferred
A metabolite of PGE2; urine levels are increased in patients with diabetic nephropathy; increased urine levels are associated with a higher risk of breast cancer in postmenopausal women with a BMI of 2
Catalog No. 50-297-0020