Learn More
Cayman Chemical tetrnor-PGJM 100ug
SDP
Supplier:  Cayman Chemical 13363100 UG
Encompass_Preferred
An expected J-ring, urinary PGD2 metabolite that may serve as a useful control in the analysis of PGD2 biosynthesis
Catalog No. 50-295-5605