Learn More
Cayman Chemical tetrnorPGDM AChE Tracer 500dtn
SDP
Supplier:  Cayman Chemical 401000500 DTN
Encompass_Preferred
An IRF3 activator and adjuvant increases the mRNA expression of the genes encoding ISG54 and OASL in PH5CH8 cells and increases CXCL10 production in phorbol 12-myristate 13-acetate-differentiated THP-1 monocytes at 10 UM immunization with a vaccine in combination with KIN1148 increases blood IgG neutralizing and hemagglutinin antibody titers decreases PFUs in the lungs and lung-draining lymph nodes and increases the survival percentage of mice infected with a lethal dose of influenza virus when used as an adjuvant
Catalog No. 50-294-3240