Learn More
Medchemexpress LLC Teucvidin | 53625-15-3 | 5 MG

Supplier: Medchemexpress LLC HYN25255MG
Teucvidin is a diterpenoid derived from Teucrium species, appearing as a white to off-white solid. Intended for research use only, it has been shown to inhibit LPS-induced NO production in mouse RAW264.7 cells with an IC50 > 50 μM.
- For research use only
- Purity of 99.67%
- Diterpenoid from Teucrium species
- Appears as a white to off-white solid
- Inhibits LPS-induced NO production in mouse RAW264.7 cells
- Recommended storage at 4°C, sealed, away from moisture and light
- In solvent storage at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.