Learn More
Medchemexpress LLC Tezacaftor | 1152311-62-0 | C26H27F3N2O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1544810MM/1ML
Encompass_Preferred
Tezacaftor is a F508del CFTR corrector that aids the CFTR protein in reaching the cell surface. When combined with Ivacaftor, a CFTR potentiator, it exhibits potent efficacy against cystic fibrosis and diseases involving homozygosity for the CFTR Phe508del mutation. The combination of Elexacaftor-Tezacaftor-Ivacaftor is intended for individuals with cystic fibrosis (CF) who have at least one Phe508del mutation, often helping them avoid the need for lung transplantation. It is suitable for laboratory research and manufacture of substances.
  • Aids CFTR protein in reaching the cell surface
  • Exhibits potent efficacy against cystic fibrosis
  • Intended for individuals with cystic fibrosis with at least one Phe508del mutation
  • Supports laboratory research
  • Used in the manufacture of substances
Catalog No. 50-003-76461