Learn More
ATCC TF-1; Erythroleukemia; Human (Homo sapiens)
Supplier:  ATCC CRL2003
Encompass
TF-1 erythroblasts are isolated from bone marrow derived from a 35-year-old Asian male with severe pancytopenia in 1987. The cells provide a good system for investigating the proliferation and differentiation of myeloid progenitor cells.
Catalog No. 50-238-2676