Learn More
ABclonal Technology TFAM Rabbit mAb
SDP
Supplier:  ABclonal Technology A23467100UL
Encompass_Preferred
This gene encodes a key mitochondrial transcription factor containing two high mobility group motifs The encoded protein also functions in mitochondrial DNA replication and repair Sequence polymorphisms in this gene are associated with Alzheimer s and Parkinson s diseases There are pseudogenes for this gene on chromosomes 6 7 and 11 Alternative splicing results in multiple transcript variants
Catalog No. 50-000-05246