Learn More
Aviva Systems Biology Corp TFCP2 Antibody - N-terminal region (ARP31073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFCP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TFCP2. Alternate names: CP2, LBP-1C, LSF, SEF, TFCP2C, LBP1C, LSF1DApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54177