Learn More
Medchemexpress LLC HY-135825 25mg , TFEB activator 1 CAS:39777-61-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13582525MG
Encompass_Preferred
HY-135825 25mg TFEB activator 1 CAS: 39777-61-2 TFEB activator 1 is an orally effective, mTOR-independent activator of TFEB. TFEB activator 1 significantly promotes the nuclear translocation of Flag-TFEB with an EC50 of 2167 nM. TFEB activator 1 enhances autophagy without inhibiting the mTOR pathway and has the potential for neurodegenerative diseases treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0775