Learn More
Cayman Chemical TFlR-NH2trIfluoroacetate s 5mg
SDP
Supplier:  Cayman Chemical 275485 MG
Encompass_Preferred
A peptide agonist of PAR1; induces calcium mobilization in isolated rat dorsal root ganglion neurons (EC50 = 1.9 μM); stimulates plasma extravasation in the bladder, esophagus, stomach, intestine, and pancreas in wild-type mice at 3 μmol/kg; reduces carrageenan-induced hyperalgesia in rats
Catalog No. 50-293-7293