Learn More
Medchemexpress LLC HY-129079 5mg Medchemexpress, TFMB-(R)-2-HG CAS:1445700-01-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1290795MG
Encompass_Preferred
Medchemexpress, HY-129079 5mg TFMB-(R)-2-HG CAS:1445700-01-5 TFMB-(R)-2-HG, a cell membrane-permeable version of (R)-2-HG, is a carcinogenic factor in Acute myeloid leukemia (AML).TFMB-(R)-2-HG impairs SCF ER-Hoxb8 cells differentiation in response to estrogen withdrawal. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9794