Learn More
Medchemexpress LLC TFMU-ADPr diammonium | 00-00-0 | 98.8% | 805.5 g/mol | C25H32F3N7O16P2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146248B5MG
Encompass_Preferred
TFMU-ADPr diammonium is a fluorescent ADP-ribose-based substrate used to monitor poly(ADP-ribose) glycohydrolase (PARG) activity in biochemical assays. Upon enzymatic cleavage the molecule releases a fluorophore, enabling quantitative measurement of PAR hydrolase activity and facilitating inhibitor screening and enzymology studies.
  • Releases fluorescent reporter upon PARG cleavage.
  • High purity (98.8%) for reproducible assay results.
  • High aqueous solubility (125 mg/mL); may require sonication.
  • Solid, white to off-white form for easy handling and storage.
  • Stable when stored under recommended conditions (-20°C under nitrogen).
Catalog No. 50-002-22641