Learn More
Medchemexpress LLC TFPI2 Human HEK293 50ug

Supplier: Medchemexpress LLC HYP71358A50UG
Recombinant human Tissue Factor Pathway Inhibitor 2 (TFPI2) expressed in HEK293 cells with a C-terminal His tag, supplied for research use. The protein corresponds to residues D23-K213 (Accession P48307) and has an approximate molecular weight of 18-32 kDa. It is characterized for protease inhibitory activity, validated in cell proliferation assays, and supplied with low endotoxin for biological assays.
- Expressed in HEK293 cells with a C-terminal His tag.
- Includes residues D23-K213 (Accession P48307).
- Approximate molecular weight 18-32 kDa.
- Endotoxin level <1 EU/μg (LAL).
- Validated bioactivity: ED50 43.15 ng/mL; specific activity 2.32x10^4 units/mg.
- Storage: store at -20°C; after reconstitution stable at 4°C for 1 week.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.