Learn More
Medchemexpress LLC TFRC Cynomolgus HE 20ug | 20ug
SDP
Supplier:  Medchemexpress LLC HYP7785520UG
Encompass_Preferred
TFRC is a key protein in cellular iron uptake coordinating receptor-mediated endocytosis of the ligand transferrin into specialized endosomes where endosomal acidification triggers iron release TFRC actively regulates T and B cell proliferation through iron uptake TFRC Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived TFRC protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-87636