Learn More
Medchemexpress LLC TG 100572 hydrochloride | 867331-64-4 | 98.8% | 512.43 g/mol | C26H27Cl2N5O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1018510MG
Encompass_Preferred
TG 100572 Hydrochloride is a multi-targeted kinase inhibitor used in research to probe VEGFR, FGFR, PDGFR, and Src family kinase signaling. It is provided as a high-purity solid suitable for biochemical and cellular assays, with recommended sealed storage away from moisture.
Catalog No. 50-002-48883