Learn More
Alkali Scientific TG 10X Solution [TRIS Glycine Buffer 10X Solution], 2 Liter
SDP
Supplier:  Alkali Scientific TGX001CP
Encompass
TG 10X Solution (Tris-Glycine Buffer) is a concentrated electrophoresis buffer commonly used in protein analysis and nucleic acid separation. The 2L solution is suitable for large-scale gel electrophoresis applications, providing a stable pH and supporting the migration of proteins and nucleic acids through an agarose or polyacrylamide gel matrix. The Tris component maintains a consistent pH, while the Glycine assists with optimal migration rates for both DNA and proteins during electrophoresis. This buffer is commonly used in academic and research laboratories for high-performance separation and analysis of biomolecules, particularly in proteomics and genomics studies.
Catalog No. 50-006-61256