Learn More
Medchemexpress LLC TG-2-in-1 (compound D003) | 135273-74-4 | 98.0% | 220.72 g/mol | C8H13ClN2OS | 50 MG

Supplier: Medchemexpress LLC HY11767850MG
TG-2-IN-1 (Compound D003) is a small-molecule inhibitor of transglutaminase-2 (TGM-2) intended for biochemical and cellular research, including studies of myopia. The compound is supplied as a high-purity research reagent with documented solubility and storage recommendations.
- High purity (98.0%).
- Molecular weight 220.72 g/mol.
- Chemical formula C8H13ClN2OS.
- Soluble in water (100 mg/mL with ultrasonic) and in DMSO (25 mg/mL with warming and ultrasonic).
- Store at 4°C, sealed, away from moisture.
- Available in multiple milligram-scale package sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.