Learn More
Apexbio Technology LLC TG003, 50mg. Cas: 300801-52-9 MFCD: MFCD00624584. Cdc2-like kinase (Clk) inhibitor
SDP
Supplier:  Apexbio Technology LLC B143150
Encompass_Preferred
TG003 is a potent and selective inhibitor of Clk-family kinases with IC50 values of 15, 20, 200 nM and >10 M for mClk4, 1, 2 and 3, respectively [1]. Also, TG003 is also an inhibitor of casein kinase 1 (CK1) [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5237
 Restricted Item