Learn More
Selleck Chemical LLC TG003 5mg 300801-52-9
SDP
Supplier:  Selleck Chemical LLC S73205MG
Encompass_Preferred
TG003 is a potent and ATP-competitive Cdc2-like kinase (Clk) inhibitor with IC50 of 20 nM, 200 nM, and 15 nM for Clk1, Clk2, and Clk4, respectively. No inhibitory effect on Clk3, SRPK1, SRPK2, or PKC. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4642