Learn More
Gold Biotechnology Inc TG1 Phage Display Electro
SDP
Supplier:  Gold Biotechnology Inc CC20510X100
Encompass_Preferred
GoldBio s TG1 Phage Display Electrocompetent E coli cells are suitable for protein expression and preparation of antibody or peptide phage display libraries TG1 Phage Display cells have 1 x 10 9 cfu/ g efficiency with electroporation and have Amber suppressor strain ( supE )
Catalog No. 50-005-62334