Learn More
TARGETMOL CHEMICALS INC TG6-129 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T289581MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. TG6-129 is an antagonist of the EP2 receptor. TG6-129 suppresses PGE2-induced elevation of cAMP in cells expressing EP2 with an IC50 value of 1.6 µM. TG6-129 reduces the expression of COX-2, IL-1beta, IL-6, IL-12, IL-23 and TNF-alpha induced by the EP2-selective agonist butaprost in P388D1 macrophages.
Catalog No. 50-287-8930