Learn More
Medchemexpress LLC TGF beta 1/TGFB1 Hu 500ug

Supplier: Medchemexpress LLC HYP7118500UG
Recombinant human transforming growth factor beta-1 (TGF-β1) expressed in CHO cells and supplied lyophilized. The protein is bioactive in multiple cell-based assays and is suitable for cell signaling studies, functional assays, and use as an assay standard. High purity and defined bioactivity support reproducible experimental results.
- Expressed in CHO cells to preserve mammalian post-translational modifications.
- Purity greater than 95% by reducing SDS-PAGE.
- Molecular weight approximately 12-14 kDa under reducing conditions.
- Potent biological activity with ED50 values in the pg-ng/mL range in multiple proliferation assays.
- Lyophilized formulation with lot-specific buffers; refer to COA for exact buffer composition.
- Stable long-term at -20°C; aliquot and store at -20°C or -80°C after reconstitution.
- Available in multiple quantities, including a 500 μg size.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.