Learn More
ABclonal Technology TGF beta induced TGFBI
SDP
Supplier:  ABclonal Technology A23275100UL
Encompass_Preferred
This gene encodes an RGD-containing protein that binds to type I II and IV collagens The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion Mutations in this gene are associated with multiple types of corneal dystrophy
Catalog No. 50-000-05315