Learn More
Medchemexpress LLC TGF-βRI inhibitor 2 | 476474-11-0 | 98.3% | 328.37 | C20H16N4O | 250 MG

Supplier: Medchemexpress LLC 250MGHYW028145
TGF-βRI inhibitor 2 is a small-molecule inhibitor of the transforming growth factor-beta receptor I (TGF-βRI) supplied for research use, primarily in cancer biology studies. The compound is delivered as a solid with a defined molecular formula and molecular weight, and is provided with high purity for in vitro and related research applications.
- Small-molecule inhibitor of transforming growth factor-beta receptor I.
- Suitable for cancer research and related cellular studies.
- High purity suitable for research use.
- Solid, light yellow to brown appearance.
- Molecular formula C20H16N4O and molecular weight 328.37.
- Recommended storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
- Available in multiple pack sizes to support different research scales.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.