Learn More
Sino Biological Peptide Substrates: TGFBR1 Peptide
Supplier:  Sino Biological T3658
Encompass_Preferred
The TGFBR1 peptide sequence (KKKVLTQMGSPSIRC-S(pS)VS) is derived from human SMAD3 (215-230) and is suitable for use as the substrate for TGFBR1 superfamily, including ACVRs (ALK1, ALK2, ALK4 and ALK7) and BMPRs (ALK3 and ALK6).
Catalog No. 50-256-7593