Learn More
Aviva Systems Biology Corp TGM3 Antibody - N-terminal region (ARP42111_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42111_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TGM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TGM3. Alternate names: MGC126249, MGC126250, TGEApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58112