Learn More
Medchemexpress LLC HY-W008574 10mg Medchemexpress, TGN-020 CAS:51987-99-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW00857410MG
Encompass_Preferred
Medchemexpress, HY-W008574 10mg TGN-020 CAS:51987-99-6 TGN-020 is a selective Aquaporin 4 (AQP4) inhibitor with an IC50 of 3.1 μM. TGN-020 is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs.TGN-020 alleviates edema and inhibits glial scar formation after spinal cord compression injury in rats[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3259