Learn More
Medchemexpress LLC TGP-377/421 (Targapre-miR-377/421) | 16752-89-9 | 98.2% | 340.38 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138647100MG
Encompass_Preferred
TGP-377/421 (Targapre-miR-377/421) is a potent miR-377 and miR-421 inhibitor that binds a functional site in both miRNAs. It is for research use only.
  • Potent miR-377 and miR-421 inhibitor
  • Binds a functional site in both miRNAs
  • Appears as a solid with a light yellow to brown color
  • Formula: C20H16N6
Catalog No. 50-003-50784