Learn More
Cayman Chemical TGR5 Receptr AgonIst 25mg
SDP
Supplier:  Cayman Chemical 1629125 MG
Encompass_Preferred
An agonist of the bile acid G protein-coupled receptor TGR5 (EC50s = 31.6-158.5 nM in cell-based assays); stimulates cAMP formation in HEK293 cells expressing TGR5 (EC50 = 2.3 pM); increases GLP-1 secretion and decreases glucose levels in the hepatic portal vein in dogs at 1 mg/kg when administered in combination with glucose; inhibits the CYP isozymes CYP2C19 and CYP3A4 (IC50s = 316.2 and 1,259 nM, respectively)
Catalog No. 50-297-2404