Learn More
Research Products International Corp TGS 10X Solution [TRIS-Glycine-SDS Buffer 10X Solution], 4 Liters
SDP
Supplier:  Research Products International Corp T320804000.0
Encompass_Preferred
Common buffer used for running SDS-PAGE gels in electrophoresis.
1X Solution Contains:
TRIS/TRIS HCl     25 mM
Glycine               0.192 M
SDS                 0.1%(w/v)
Catalog No. 50-489-084