Learn More
TARGETMOL CHEMICALS INC TGX-221 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T266110MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TGX-221 an effective specific and cell membrane permeable inhibitor of the PI3K p110(beta) catalytic subunit is utilized for cancer treatment. purity: 99%
Catalog No. 50-257-7068