Learn More
Selleck Chemical LLC TGX-221 25mg 663619-89-4
SDP
Supplier:  Selleck Chemical LLC S116925MG
Encompass_Preferred
TGX-221 is a p110?-specific inhibitor with IC50 of 5 nM in a cell-free assay, 1000-fold more selective for p110? than p110?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-865-8