Learn More
Genetex Inc Tyrosine Hydroxylase Monoclonal Antibody [HMV312] HistoMAX™ (GTX639936) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639936
Encompass_Preferred
The Tyrosine Hydroxylase antibody [HMV312] HistoMAX™ (GTX639936) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human heart muscle tissue using GTX639936 Tyrosine Hydroxylase antibody [HMV312] HistoMAX. Strong tyrosine hydroxylase staining of fibers in a small nerve.
Catalog No. NC3669311