Learn More
Medchemexpress LLC TH-Z93 | 2260887-09-8 | C12H22N2O7P2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY150167
Encompass_Preferred
TH-Z93 is a lipophilic bisphosphonate and an FPPS inhibitor with an IC50 of 90 nM. It has shown strong prophylactic effects in a pathogenic influenza model and inhibited tumor growth in a B16-OVA cells xenograft mice model, making it suitable for various research applications.
  • Lipophilic bisphosphonate
  • FPPS inhibitor (IC50: 90 nM)
  • Shows strong prophylactic effects in pathogenic influenza model
  • Inhibits tumor growth in B16-OVA cells xenograft mice model
  • High purity (98.41%)
  • Available in various package sizes
Catalog No. 50-003-74498