Learn More
Medchemexpress LLC HY-120574 5mg Medchemexpress, TH1338 CAS:1258494-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1205745MG
Encompass_Preferred
Medchemexpress, HY-120574 5mg TH1338 CAS:1258494-60-8 TH1338 (compound 3b), an orally active camptothecin derivative and a potent chemotherapeutic agent for cancer, demonstrates excellent cytotoxic potency against human tumor cell lines in vitro. TH1338 (compound 3b) possesses significant brain penetration, favorable efflux pump properties, and hematological toxicity profile. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0942