Learn More
Medchemexpress LLC TH34 | 2196203-96-8 | 99.5% | 256.30 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11181810MM/1ML
Encompass_Preferred
TH34 is an inhibitor of HDAC6, HDAC8, and HDAC10, with IC50 values of 4.6 μM, 1.9 μM, and 7.7 μM, respectively. It exhibits high selectivity over HDAC1, HDAC2, and HDAC3. For research use only, not intended for patients. TH34 induces caspase-dependent programmed cell death and cell cycle arrest in neuroblastoma cells.
  • HDAC6/8/10 inhibitor
  • IC50s: 4.6 μM (HDAC6), 1.9 μM (HDAC8), 7.7 μM (HDAC10)
  • High selectivity over HDAC1/2/3
  • Induces caspase-dependent programmed cell death in neuroblastoma cells
  • Induces cell cycle arrest in neuroblastoma cells
  • Appearance: solid
  • Color: white to off-white
  • For research use only
Catalog No. 50-004-39288