Learn More
Selleck Chemical LLC TH34 S8773-5MG
SDP
Supplier:  Selleck Chemical LLC S87735MG
Encompass_Preferred
TH34 is a HDAC inhibitor that shows pronounced selectivity for HDACs 6 8 and 10 over HDACs 1 2 and 3 In a NanoBRET assay TH34 strongly binds HDAC6 8 and 10 with low-micromolar IC50 concentrations (HDAC6 4 6 M HDAC8 1 9 M HDAC10 7 7 M)
Catalog No. 50-000-24098