Learn More
Selleck Chemical LLC TH5427 E2849-25MG
SDP
Supplier:  Selleck Chemical LLC E284925MG
Encompass_Preferred
TH5427 is a potent and cell-active NUDT5 (also called NUDIX5) inhibitor that can be used to further understand the role of NUDT5 in biological systems
Catalog No. 50-000-40572