Learn More
TARGETMOL CHEMICALS INC TH5487 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T811910MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TH5487 is an potent inhibitor of 8-oxoguanine DNA glycosylase 1 (OGG1)( IC50 of 342 nM)purity: 98%
Catalog No. 50-257-9101