Learn More
Cayman Chemical THAL-SNS-032 10mg
SDP
Supplier:  Cayman Chemical 4095810 MG
Encompass_Preferred
A PROTAC that drives Cdk9 degradation; selectively induces degradation of Cdk9 over Cdk1, Cdk2, and Cdk7 in MOLT-4 cells at 250 nM; inhibits HCMV replication in infected HFFs (EC50s = 0.025 µM); inhibits viral replication in murine CMV-infected MEFs (EC50 = 0.22 µM)
Catalog No. 50-294-2791