Learn More
Medchemexpress LLC Thalidomide-1-Me,4-OH | 2244568-06-5 | >98.0% | 288.26 | C14H12N2O5 | 25 MG

Supplier: Medchemexpress LLC HYW44137625MG
Thalidomide-1-Me,4-OH is a thalidomide analogue with a 1-methyl and 4-hydroxyl substitution that functions as an E3 ubiquitin ligase ligand. It is used in biochemical research to recruit E3 ligases (Cereblon-related activity) for targeted protein degradation studies.
- Acts as an E3 ligase ligand suitable for cereblon-mediated recruitment.
- Contains 1-methyl and 4-hydroxyl substitutions on the thalidomide scaffold.
- Molecular formula C14H12N2O5, molecular weight 288.26.
- Purity greater than 98%.
- Recommended storage: powder at -20°C (long-term) or 4°C (short-term); in solvent at -80°C for extended storage.
- Supplied in small research quantities, e.g., 25 MG, for biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.